siRNA Details
| Gene Symbol: Hirip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025725.1 | NP_001020896.1 | 8 | 2057 | ||
| XM_006230307.1 | XP_006230369.1 | 7 | 1754 | |||
| GenBank mRNA | BC099091.1 | - | 8 | 2057 | 7457888 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hirip3| Gene Symbol: Hirip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025725.1 | NP_001020896.1 | 8 | 2057 | ||
| XM_006230307.1 | XP_006230369.1 | 7 | 1754 | |||
| GenBank mRNA | BC099091.1 | - | 8 | 2057 | 7457888 | |