siRNA Details
| Gene Symbol: Rnf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025727.1 | NP_001020898.1 | 2 | 305 | ||
| XM_006256225.2 | XP_006256287.1 | 2 | 382 | |||
| XM_006256226.2 | XP_006256288.1 | 2 | 385 | |||
| GenBank mRNA | BC099079.1 | - | 2 | 305 | 7443362 | |

Rnf8
Genomic Map