siRNA Details
| Gene Symbol: Zbed3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025729.1 | NP_001020900.1 | 2 | 752 | ||
| XM_006231816.2 | XP_006231878.1 | 2 | 894 | |||
| GenBank mRNA | BC097974.1 | - | 2 | 752 | 7442124 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbed3| Gene Symbol: Zbed3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025729.1 | NP_001020900.1 | 2 | 752 | ||
| XM_006231816.2 | XP_006231878.1 | 2 | 894 | |||
| GenBank mRNA | BC097974.1 | - | 2 | 752 | 7442124 | |