siRNA Details
| Gene Symbol: Gle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025731.1 | NP_001020902.1 | 10 | 1386 | ||
| XM_006233836.2 | XP_006233898.1 | 10 | 1422 | |||
| XM_006233838.2 | XP_006233900.1 | 10 | 1421 | |||
| GenBank mRNA | BC099088.1 | - | 10 | 1386 | 7456179 | |
| FQ217346.1 | - | 10 | 1407 | |||

Gle1
Genomic Map