siRNA Details
| Gene Symbol: Nop56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025732.1 | NP_001020903.1 | 12 | 1641 | ||
| GenBank mRNA | BC086568.1 | - | 9 | 1319 | ||
| BC099149.1 | - | 12 | 1641 | 7366316 | ||
| FQ232640.1 | - | 7 | 887 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nop56| Gene Symbol: Nop56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025732.1 | NP_001020903.1 | 12 | 1641 | ||
| GenBank mRNA | BC086568.1 | - | 9 | 1319 | ||
| BC099149.1 | - | 12 | 1641 | 7366316 | ||
| FQ232640.1 | - | 7 | 887 | |||