siRNA Details
| Gene Symbol: Nop56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025732.1 | NP_001020903.1 | 10 | 1307 | ||
| GenBank mRNA | BC086568.1 | - | 7 | 985 | ||
| BC099149.1 | - | 10 | 1307 | 7366316 | ||
| FQ224806.1 | - | 2 | 560 | |||
| FQ232640.1 | - | 5 | 553 | |||

Nop56
Genomic Map