siRNA Details
| Gene Symbol: Tcea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025735.1 | NP_001020906.1 | 10 | 1181 | ||
| XM_008763546.1 | XP_008761768.1 | 10 | 1069 | |||
| GenBank mRNA | BC099141.1 | - | 10 | 1181 | 7460791 | |
| FQ227501.1 | - | 10 | 1153 | |||

Tcea1
Genomic Map