siRNA Details
| Gene Symbol: Gnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025736.1 | NP_001020907.1 | 8 | 942 | ||
| GenBank mRNA | BC099173.1 | - | 8 | 942 | 7382617 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnl2| Gene Symbol: Gnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025736.1 | NP_001020907.1 | 8 | 942 | ||
| GenBank mRNA | BC099173.1 | - | 8 | 942 | 7382617 | |