siRNA Details
| Gene Symbol: Eml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025741.1 | NP_001020912.1 | 1243 | |||
| GenBank mRNA | BC097450.1 | - | 1243 | 7456735 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eml1| Gene Symbol: Eml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025741.1 | NP_001020912.1 | 1243 | |||
| GenBank mRNA | BC097450.1 | - | 1243 | 7456735 | ||