siRNA Details
| Gene Symbol: Dazap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025742.1 | NP_001020913.1 | 5 | 465 | ||
| XM_006240968.2 | XP_006241030.1 | 5 | 532 | |||
| XM_006240969.2 | XP_006241031.1 | 5 | 414 | |||
| XM_006240970.2 | XP_006241032.1 | 5 | 411 | |||
| XM_008765123.1 | XP_008763345.1 | 5 | 1040 | |||
| GenBank mRNA | BC083601.1 | - | 5 | 437 | ||
| BC098930.1 | - | 5 | 465 | 7133199 | ||

Dazap1
Genomic Map