siRNA Details
| Gene Symbol: Zfand2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025745.1 | NP_001020916.1 | 9 | 1122 | ||
| XM_006245245.2 | XP_006245307.1 | 10 | 928 | |||
| XM_006245246.2 | XP_006245308.1 | 10 | 910 | |||
| GenBank mRNA | BC099215.1 | - | 9 | 1122 | 7384656 | |

Zfand2b
Genomic Map