siRNA Details
| Gene Symbol: Taz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025748.1 | NP_001020919.1 | 10 | 1397 | ||
| XM_006229599.2 | XP_006229661.1 | 11 | 1788 | |||
| XM_006229600.2 | XP_006229662.1 | 12 | 1579 | |||
| GenBank mRNA | BC099221.1 | - | 10 | 1397 | 7387882 | |

Taz
Genomic Map