siRNA Details
| Gene Symbol: Rem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025753.1 | NP_001020924.1 | 5 | 1197 | ||
| XM_006235278.2 | XP_006235340.1 | 5 | 1339 | |||
| GenBank mRNA | BC098946.1 | - | 5 | 1197 | 7460067 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rem1| Gene Symbol: Rem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025753.1 | NP_001020924.1 | 5 | 1197 | ||
| XM_006235278.2 | XP_006235340.1 | 5 | 1339 | |||
| GenBank mRNA | BC098946.1 | - | 5 | 1197 | 7460067 | |