siRNA Details
| Gene Symbol: Tmem186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025756.1 | NP_001020927.1 | 2 | 1758 | ||
| GenBank mRNA | BC098933.1 | - | 2 | 1758 | 7386881 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem186| Gene Symbol: Tmem186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025756.1 | NP_001020927.1 | 2 | 1758 | ||
| GenBank mRNA | BC098933.1 | - | 2 | 1758 | 7386881 | |