siRNA Details
| Gene Symbol: MGC112692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025759.1 | NP_001020930.1 | 683 | |||
| XM_008768997.1 | XP_008767219.1 | 2 | 719 | |||
| GenBank mRNA | BC081909.1 | - | 679 | |||
| BC098706.1 | - | 683 | 7114401 | |||

MGC112692
Genomic Map