siRNA Details
| Gene Symbol: LOC498933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025764.1 | NP_001020935.1 | 6 | 784 | ||
| GenBank mRNA | BC098038.1 | - | 6 | 784 | 7456623 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC498933| Gene Symbol: LOC498933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025764.1 | NP_001020935.1 | 6 | 784 | ||
| GenBank mRNA | BC098038.1 | - | 6 | 784 | 7456623 | |