siRNA Details
| Gene Symbol: Spata25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025769.1 | NP_001020940.1 | 2 | 600 | ||
| GenBank mRNA | BC097436.1 | - | 2 | 600 | 7455517 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata25| Gene Symbol: Spata25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025769.1 | NP_001020940.1 | 2 | 600 | ||
| GenBank mRNA | BC097436.1 | - | 2 | 600 | 7455517 | |