siRNA Details
| Gene Symbol: Fam221a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025771.2 | NP_001020942.1 | 4 | 504 | ||
| XM_006236467.2 | XP_006236529.1 | 4 | 477 | |||
| XM_006236468.2 | XP_006236530.1 | 3 | 491 | |||
| GenBank mRNA | BC097434.1 | - | 4 | 505 | 7455430 | |

Fam221a
Genomic Map