siRNA Details
| Gene Symbol: Stpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025772.1 | NP_001020943.1 | 9 | 1416 | ||
| GenBank mRNA | BC083648.1 | - | 8 | 1358 | ||
| BC098937.1 | - | 9 | 1416 | 7453388 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stpg1| Gene Symbol: Stpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025772.1 | NP_001020943.1 | 9 | 1416 | ||
| GenBank mRNA | BC083648.1 | - | 8 | 1358 | ||
| BC098937.1 | - | 9 | 1416 | 7453388 | ||