siRNA Details
| Gene Symbol: Tnfrsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025773.1 | NP_001020944.1 | 6 | 864 | ||
| XM_006239471.2 | XP_006239533.1 | 7 | 1284 | |||
| XM_006239472.2 | XP_006239534.1 | 6 | 1717 | |||
| XM_008764282.1 | XP_008762504.1 | 6 | 1054 | |||
| XM_008764283.1 | XP_008762505.1 | 7 | 1305 | |||
| GenBank mRNA | AB164643.1 | - | 5 | 440 | ||
| BC097483.1 | - | 6 | 864 | 7459228 | ||

Tnfrsf9
Genomic Map