siRNA Details
| Gene Symbol: Ppip5k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025775.1 | NP_001020946.1 | 10, 11 | 1396 | ||
| GenBank mRNA | BC097365.1 | - | 10, 11 | 1396 | 7443073 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppip5k2| Gene Symbol: Ppip5k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025775.1 | NP_001020946.1 | 10, 11 | 1396 | ||
| GenBank mRNA | BC097365.1 | - | 10, 11 | 1396 | 7443073 | |