siRNA Details
| Gene Symbol: Actr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031068.1 | NP_112330.1 | 12 | 1409 | ||
| GenBank mRNA | BC098014.1 | - | 12 | 1409 | 7454624 | |
| FQ212256.1 | - | 12 | 1611 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actr3| Gene Symbol: Actr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031068.1 | NP_112330.1 | 12 | 1409 | ||
| GenBank mRNA | BC098014.1 | - | 12 | 1409 | 7454624 | |
| FQ212256.1 | - | 12 | 1611 | |||