siRNA Details
| Gene Symbol: Strn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240075.2 | XP_006240137.1 | 1, 2 | 578 | ||
| XM_006240076.2 | XP_006240138.1 | 1, 2 | 577 | |||
| XM_006240077.2 | XP_006240139.1 | 1, 2 | 566 | |||
| GenBank mRNA | AY026526.1 | - | 1, 2 | 505 | ||
| DQ473607.1 | - | 1, 2 | 460 | |||

Strn3
Genomic Map