siRNA Details
| Gene Symbol: Tjp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053773.1 | NP_446225.1 | 3, 4 | 353 | ||
| XM_006231175.2 | XP_006231237.1 | 3, 4 | 1435 | |||
| XM_006231176.2 | XP_006231238.1 | 3, 4 | 1436 | |||
| XM_006231177.2 | XP_006231239.1 | 2, 3 | 666 | |||
| XM_006231178.2 | XP_006231240.1 | 2, 3 | 3595 | |||
| XM_006231179.2 | XP_006231241.1 | 2, 3 | 499 | |||
| XM_006231181.2 | XP_006231243.1 | 3, 4 | 454 | |||
| XM_006231182.2 | XP_006231244.1 | 2, 3 | 666 | |||
| XM_008760302.1 | XP_008758524.1 | 3, 4 | 195 | |||
| GenBank mRNA | BC103479.1 | - | 3, 4 | 353 | 7936785 | |

Tjp2
Genomic Map