siRNA Details
| Gene Symbol: Tjp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053773.1 | NP_446225.1 | 5 | 531 | ||
| XM_006231175.2 | XP_006231237.1 | 5 | 1613 | |||
| XM_006231176.2 | XP_006231238.1 | 5 | 1614 | |||
| XM_006231177.2 | XP_006231239.1 | 4 | 844 | |||
| XM_006231178.2 | XP_006231240.1 | 4 | 3773 | |||
| XM_006231179.2 | XP_006231241.1 | 4 | 677 | |||
| XM_006231181.2 | XP_006231243.1 | 5 | 632 | |||
| XM_006231182.2 | XP_006231244.1 | 4 | 844 | |||
| XM_008760302.1 | XP_008758524.1 | 5 | 373 | |||
| GenBank mRNA | BC103479.1 | - | 5 | 531 | 7936785 | |

Tjp2
Genomic Map