siRNA Details
| Gene Symbol: Kif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057202.1 | NP_476550.1 | 10 | 895 | ||
| XM_006254005.1 | XP_006254067.1 | 10 | 1326 | |||
| GenBank mRNA | DQ309275.1 | - | 10 | 895 | ||
| FQ226753.1 | - | 10 | 1289 | |||
| FQ227346.1 | - | 8 | 686 | |||
| FQ230391.1 | - | 10 | 1318 | |||
| FQ232221.1 | - | 8 | 686 | |||
| FQ233216.1 | - | 10 | 1300 | |||
| FQ234099.1 | - | 8 | 686 | |||
| FQ234494.1 | - | 8 | 686 | |||
| FQ235192.1 | - | 8 | 686 | |||

Kif5b
Genomic Map