siRNA Details
| Gene Symbol: Kif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057202.1 | NP_476550.1 | 2 | 140 | ||
| XM_006254005.1 | XP_006254067.1 | 2 | 571 | |||
| GenBank mRNA | DQ309275.1 | - | 2 | 140 | ||
| FQ226753.1 | - | 2 | 534 | |||
| FQ230391.1 | - | 2 | 563 | |||
| FQ233216.1 | - | 2 | 543 | |||

Kif5b
Genomic Map