siRNA Details
| Gene Symbol: Cox8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134345.1 | NP_599172.1 | 2 | 440 | ||
| GenBank mRNA | BC126066.1 | - | 2 | 407 | 7385046 | |
| L48209.1 | - | 2 | 358 | |||
| M28255.1 | - | 1 | 279 | |||
| X06146.1 | - | 285 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cox8a| Gene Symbol: Cox8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134345.1 | NP_599172.1 | 2 | 440 | ||
| GenBank mRNA | BC126066.1 | - | 2 | 407 | 7385046 | |
| L48209.1 | - | 2 | 358 | |||
| M28255.1 | - | 1 | 279 | |||
| X06146.1 | - | 285 | ||||