siRNA Details
| Gene Symbol: C21orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162495.2 | NP_001155967.2 | 4 | 1098 | ||
| NM_001162496.2 | NP_001155968.2 | 2 | 942 | |||
| NM_019596.5 | NP_062542.5 | 3 | 1023 | |||
| GenBank mRNA | AF231922.1 | - | 3 | 928 | ||

C21orf62
Genomic Map