siRNA Details
| Gene Symbol: Syt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765352.1 | XP_008763574.1 | 8 | 3709 | ||
| XM_008765353.1 | XP_008763575.1 | 8 | 3703 | |||
| GenBank mRNA | X52772.1 | - | 11 | 3906 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syt1| Gene Symbol: Syt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765352.1 | XP_008763574.1 | 8 | 3709 | ||
| XM_008765353.1 | XP_008763575.1 | 8 | 3703 | |||
| GenBank mRNA | X52772.1 | - | 11 | 3906 | ||