siRNA Details
| Gene Symbol: Rpain | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033060.1 | NP_001028232.1 | 2 | 206 | ||
| XM_006246668.2 | XP_006246730.1 | 2 | 570 | |||
| GenBank mRNA | AY775322.1 | - | 2 | 206 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpain| Gene Symbol: Rpain | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033060.1 | NP_001028232.1 | 2 | 206 | ||
| XM_006246668.2 | XP_006246730.1 | 2 | 570 | |||
| GenBank mRNA | AY775322.1 | - | 2 | 206 | ||