siRNA Details
| Gene Symbol: SUSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019601.3 | NP_062547.1 | 15 | 3105 | ||
| GenBank mRNA | AK126105.1 | - | 11 | 4860 | ||
| BC033107.1 | - | 15 | 3105 | 5457186 | ||
| BI822588.1 | - | 261 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SUSD2| Gene Symbol: SUSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019601.3 | NP_062547.1 | 15 | 3105 | ||
| GenBank mRNA | AK126105.1 | - | 11 | 4860 | ||
| BC033107.1 | - | 15 | 3105 | 5457186 | ||
| BI822588.1 | - | 261 | ||||