siRNA Details
Gene Symbol: Shpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033682.1 | NP_001028854.1 | 7 | 1452 | ||
GenBank mRNA | BC101914.1 | - | 7 | 1452 | 7935768 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Shpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033682.1 | NP_001028854.1 | 7 | 1452 | ||
GenBank mRNA | BC101914.1 | - | 7 | 1452 | 7935768 |