siRNA Details
| Gene Symbol: Blmh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034163.1 | NP_001029335.1 | 12 | 1696 | ||
| GenBank mRNA | BC128701.1 | - | 12 | 1581 | 7379958 | |
| D87336.1 | - | 12 | 1600 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Blmh| Gene Symbol: Blmh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034163.1 | NP_001029335.1 | 12 | 1696 | ||
| GenBank mRNA | BC128701.1 | - | 12 | 1581 | 7379958 | |
| D87336.1 | - | 12 | 1600 | |||