siRNA Details
| Gene Symbol: Vat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033683.1 | NP_001028855.1 | 5 | 1093 | ||
| GenBank mRNA | BC101882.1 | - | 5 | 1093 | 7929680 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vat1| Gene Symbol: Vat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033683.1 | NP_001028855.1 | 5 | 1093 | ||
| GenBank mRNA | BC101882.1 | - | 5 | 1093 | 7929680 | |