siRNA Details
| Gene Symbol: Pyy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034080.1 | NP_001029252.1 | 4 | 412 | ||
| GenBank mRNA | AB238226.1 | - | 4 | 412 | ||
| M17523.1 | - | 3 | 391 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pyy| Gene Symbol: Pyy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034080.1 | NP_001029252.1 | 4 | 412 | ||
| GenBank mRNA | AB238226.1 | - | 4 | 412 | ||
| M17523.1 | - | 3 | 391 | |||