siRNA Details
| Gene Symbol: Psmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031639.1 | NP_001026809.1 | 8 | 1065 | ||
| GenBank mRNA | BC099135.1 | - | 8 | 1065 | 7455692 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmd2| Gene Symbol: Psmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031639.1 | NP_001026809.1 | 8 | 1065 | ||
| GenBank mRNA | BC099135.1 | - | 8 | 1065 | 7455692 | |