siRNA Details
| Gene Symbol: Nit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034126.1 | NP_001029298.1 | 4, 5 | 378 | ||
| GenBank mRNA | BC082034.1 | - | 4, 5 | 378 | ||
| BC100637.1 | - | 4, 5 | 378 | 5621093 | ||
| FQ209436.1 | - | 4, 5 | 386 | |||
| FQ219054.1 | - | 1, 2 | 310 | |||
| FQ219183.1 | - | 4, 5 | 384 | |||
| FQ229934.1 | - | 4, 5 | 390 | |||

Nit2
Genomic Map