siRNA Details
| Gene Symbol: Nptx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034199.1 | NP_001029371.1 | 1 | 253 | ||
| GenBank mRNA | CB578587.1 | - | 1 | 258 | ||
| CO403402.1 | - | 1 | 215 | |||
| S82649.1 | - | 187 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nptx2| Gene Symbol: Nptx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034199.1 | NP_001029371.1 | 1 | 253 | ||
| GenBank mRNA | CB578587.1 | - | 1 | 258 | ||
| CO403402.1 | - | 1 | 215 | |||
| S82649.1 | - | 187 | ||||