siRNA Details
| Gene Symbol: Eif3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031640.1 | NP_001026810.1 | 8 | 1280 | ||
| GenBank mRNA | BC098728.1 | - | 8 | 1280 | 7381504 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_133916, BC051065, AK219469, AK089978, AK198886, and others.

Eif3b
Genomic Map