siRNA Details
| Gene Symbol: Ifi30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030026.1 | NP_001025197.1 | 5, 6 | 766 | ||
| GenBank mRNA | BC088256.1 | - | 5, 6 | 630 | ||
| BC099774.1 | - | 5, 6 | 766 | 7932222 | ||
| FQ225096.1 | - | 5, 6 | 652 | |||
| FQ229307.1 | - | 5, 6 | 806 | |||
| FQ230297.1 | - | 5, 6 | 653 | |||
| FQ232607.1 | - | 5, 6 | 652 | |||
| FQ233058.1 | - | 5, 6 | 652 | |||
| FQ233214.1 | - | 5, 6 | 652 | |||
| FQ234315.1 | - | 5, 6 | 652 | |||

Ifi30
Genomic Map