siRNA Details
| Gene Symbol: LOC290876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037182.1 | NP_001032259.1 | 8, 9 | 930 | ||
| XM_006253430.2 | XP_006253492.1 | 8, 9 | 1139 | |||
| XM_006253433.2 | XP_006253495.1 | 8, 9 | 934 | |||
| XM_008771386.1 | XP_008769608.1 | 8, 9 | 1178 | |||
| GenBank mRNA | AB714639.1 | - | 2, 3 | 163 | ||
| BC085810.1 | - | 8, 9 | 890 | |||
| BC100249.1 | - | 8, 9 | 930 | 7135908 | ||

LOC290876
Genomic Map