siRNA Details
| Gene Symbol: Nkapl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029913.1 | NP_001025084.1 | 1 | 1351 | ||
| GenBank mRNA | BC097473.1 | - | 1 | 1351 | 7457806 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkapl| Gene Symbol: Nkapl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029913.1 | NP_001025084.1 | 1 | 1351 | ||
| GenBank mRNA | BC097473.1 | - | 1 | 1351 | 7457806 | |