siRNA Details
| Gene Symbol: Ercc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031644.1 | NP_001026814.1 | 2 | 250 | ||
| GenBank mRNA | BC098856.1 | - | 2 | 250 | 7379944 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ercc3| Gene Symbol: Ercc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031644.1 | NP_001026814.1 | 2 | 250 | ||
| GenBank mRNA | BC098856.1 | - | 2 | 250 | 7379944 | |