siRNA Details
| Gene Symbol: Iws1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034918.1 | NP_001030090.1 | 10 | 2100 | ||
| XM_006254571.2 | XP_006254633.1 | 10 | 2131 | |||
| XM_006254572.2 | XP_006254634.1 | 10 | 2131 | |||
| XM_006254573.2 | XP_006254635.1 | 10 | 2128 | |||
| XM_006254574.2 | XP_006254636.1 | 10 | 2131 | |||
| XM_008772034.1 | XP_008770256.1 | 10 | 2131 | |||
| GenBank mRNA | BC104700.1 | - | 10 | 2100 | 7190021 | |
| FQ227776.1 | - | 2 | 211 | |||

Iws1
Genomic Map