siRNA Details
| Gene Symbol: Cmtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029914.1 | NP_001025085.1 | 4 | 1296 | ||
| XM_006255052.2 | XP_006255114.1 | 3 | 1187 | |||
| GenBank mRNA | BC098029.1 | - | 4 | 1296 | 7456234 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmtm1| Gene Symbol: Cmtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029914.1 | NP_001025085.1 | 4 | 1296 | ||
| XM_006255052.2 | XP_006255114.1 | 3 | 1187 | |||
| GenBank mRNA | BC098029.1 | - | 4 | 1296 | 7456234 | |