siRNA Details
| Gene Symbol: Cmtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029914.1 | NP_001025085.1 | 1 | 818 | ||
| XM_006255052.2 | XP_006255114.1 | 1 | 868 | |||
| GenBank mRNA | BC098029.1 | - | 1 | 818 | 7456234 | |
| Gene Symbol: Cklf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772240.1 | XP_008770462.1 | 3 | 1850 | ||

Cmtm1
Genomic Map