siRNA Details
| Gene Symbol: Acvr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031571.2 | NP_113759.1 | 2 | 198 | ||
| XM_008761703.1 | XP_008759925.1 | 2 | 253 | |||
| GenBank mRNA | L10639.1 | - | 2 | 181 | ||
| S48190.1 | - | 2 | 198 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acvr2a| Gene Symbol: Acvr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031571.2 | NP_113759.1 | 2 | 198 | ||
| XM_008761703.1 | XP_008759925.1 | 2 | 253 | |||
| GenBank mRNA | L10639.1 | - | 2 | 181 | ||
| S48190.1 | - | 2 | 198 | |||