siRNA Details
| Gene Symbol: Numbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033888.1 | NP_001029060.1 | 2 | 43 | ||
| XM_006228565.2 | XP_006228627.1 | 1 | 3207 | |||
| XM_006228566.2 | XP_006228628.1 | 1 | 3207 | |||
| XM_006228567.2 | XP_006228629.1 | 2 | 266 | |||
| XM_006228568.2 | XP_006228630.1 | 2 | 472 | |||
| GenBank mRNA | AB210107.1 | - | 2 | 43 | ||
| BC129073.1 | - | 2 | 153 | 7189989 | ||

Numbl
Genomic Map