siRNA Details
| Gene Symbol: Plcb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033350.1 | NP_203501.1 | 9 | 860 | ||
| XM_006230711.2 | XP_006230773.1 | 9 | 992 | |||
| GenBank mRNA | DQ120507.1 | - | 9 | 860 | ||
| DQ120508.1 | - | 9 | 860 | |||
| M99567.1 | - | 9 | 794 | |||

Plcb3
Genomic Map